Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
bcl-x Antibody (5E9), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | bcl-x |
---|---|
Clone | 5E9 |
Dilution | Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Classification | Monoclonal |
Description
bcl-x Monoclonal antibody specifically detects bcl-x in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
bcl-x | |
Western Blot 1:500-1:5000, ELISA, Immunohistochemistry 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
Apoptosis regulator Bcl-X,BCL2L,BCL2L1,Bcl-2-like protein 1,BCLX,Bcl-X,BCL-XL/S,PPP1R52 | |
A synthesized peptide derived from Human Bcl-x [UniProt Q07817] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
5E9 | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Apoptosis, Biologically Active Proteins, Cancer, Phospho Specific, Tumor Suppressors | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
598 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title