Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-xL Antibody (7B2.5), Biotin, Novus Biologicals™
SDP

Catalog No. NBP128561 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP128561 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP128561 Supplier Novus Biologicals Supplier No. NBP128561
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Bcl-xL Monoclonal specifically detects Bcl-xL in Human, Mouse, Rat, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Bcl-xL
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 7B2.5
Concentration 0.1mg/mL
Conjugate Biotin
Dilution Western Blot 1:100-1:2000, Flow Cytometry < = 3 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Gene Alias Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092
Gene Symbols BCL2L1
Host Species Mouse
Immunogen Recombinant human Bcl-xS
Purification Method Protein A or G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 598
Test Specificity Human/Mouse/Rat/Rhesus Bcl-xL
Target Species Human, Mouse, Rat, Rhesus Macaque
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG3
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.