Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-xL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11083982
Description
Bcl-xL Polyclonal specifically detects Bcl-xL in Human, Canine, Feline, Sheep samples. It is validated for Western Blot, ICC/IF (Negative).Specifications
| Bcl-xL | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
| Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
| Rabbit | |
| 26 kDa | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 598 | |
| Human, Canine, Feline, Sheep | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| Western Blot 2 ug/ml, ICC/IF (Negative) | |
| Q07817 | |
| BCL2L1 | |
| Synthetic peptide made to an N-terminal portion of human BCL2L1 (within residues 30-70). [Swiss-Prot# Q07817] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| There are three known isoforms of Bcl-X; Bcl-X(S), Bcl-X(beta), and Bcl-X(L). The immunogen used to create this has 100% identity to all three isoforms. | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction