Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-xL Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Specifications
| Antigen | Bcl-xL |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 ug/ml, ICC/IF (Negative) |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB11083982
![]() |
Novus Biologicals
NB11083982 |
0.1 mL | N/A | N/A | N/A | ||||
Description
Bcl-xL Polyclonal specifically detects Bcl-xL in Human, Canine, Feline, Sheep samples. It is validated for Western Blot, ICC/IF (Negative).Specifications
| Bcl-xL | |
| Western Blot 2 ug/ml, ICC/IF (Negative) | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, Cancer, Tumor Suppressors | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.1% Sodium Azide | |
| Apoptosis regulator Bcl-X, bcl2-L-1, BCL2-like 1, bcl-2-like protein 1, BCLXBCL2LBcl-X, bcl-xL, BCLXL, BCL-XL/S, bcl-xS, BCLXS, DKFZp781P2092 | |
| BCL2L1 | |
| IgG | |
| Affinity Purified | |
| There are three known isoforms of Bcl-X; Bcl-X(S), Bcl-X(beta), and Bcl-X(L). The immunogen used to create this has 100% identity to all three isoforms. |
| 1 mg/mL | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Human, Canine, Feline, Sheep | |
| Q07817 | |
| 598 | |
| Synthetic peptide made to an N-terminal portion of human BCL2L1 (within residues 30-70). [Swiss-Prot# Q07817] | |
| Primary | |
| Store at 4C. Do not freeze. | |
| 26 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title