Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCL11A Monoclonal Antibody (BCL11A/123), Active Motif™

Mouse Monoclonal Antibody
$156.65 - $620.58
Specifications
Antigen | BCL11A |
---|---|
Clone | BCL11A/123 |
Concentration | 1 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3253
![]() ![]() |
Active Motif
61232 |
10 μg |
Each for $156.65
|
|
|||||
50-199-3252
![]() ![]() |
Active Motif
61231 |
100 μg |
Each for $620.58
|
|
|||||
Description
This gene encodes a C2H2 type zinc-finger protein by its similarity to the mouse Bcl11a/Evi9 protein. The corresponding mouse gene is a common site of retroviral integration in myeloid leukemia, and may function as a leukemia disease gene, in part, through its interaction with BCL6. During hematopoietic cell differentiation, this gene is down-regulated. It is possibly involved in lymphoma pathogenesis since translocations associated with B-cell malignancies also deregulates its expression. Multiple transcript variants encoding several different isoforms have been found for this gene.Specifications
BCL11A | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
2810047E18Rik; B cell CLL/lymphoma 11A (zinc finger protein); B-cell CLL/lymphoma 11A; B-cell CLL/lymphoma 11A (zinc finger protein); B-cell CLL/lymphoma 11A (zinc finger protein) isoform 2; B-cell lymphoma/leukemia 11A; Bcl11a; BCL-11A; BCL11A B-cell CLL/lymphoma 11A (zinc finger protein) isoform 1; BCL11A, BAF complex component; BCL11A-L; BCL11a-M; BCL11A-S; BCL11A-XL; BOS_11500; C2H2-type zinc finger protein; COUP-TF interacting protein 1; COUP-TF-interacting protein 1; Ctip1; D930021L15Rik; ecotropic viral integration site 9 homolog; Ecotropic viral integration site 9 protein; Ecotropic viral integration site 9 protein homolog; EVI9; EVI-9; Evi9a; Evi9b; Evi9c; FLJ10173; FLJ34997; HBFQTL5; KIAA1809; mKIAA1809; myeloid leukemia; Zinc finger protein 856; ZNF856 | |
BCL11A | |
IgG1 | |
Protein G | |
Antibody |
BCL11A/123 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9H165 | |
53335 | |
This BCL11A antibody was raised against a recombinant protein corresponding to amino acids 637-835 of BCL11A-XL (B-cell lymphoma/leukemia 11A extra long form). | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles | |
BCL11A |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title