Learn More
BCL11A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 116131AP150UL

Description
This is a rabbit polyclonal antibody raised against part chain of N-terminal BCL11A of human origin. There are three isoforms of BCL11A: isoform1 (773 aa), isoform2 (486 aa), and isoform3 (239 aa).
This gene encodes a C2H2 type zinc-finger protein by its similarity to the mouse Bcl11a/Evi9 protein. The corresponding mouse gene is a common site of retroviral integration in myeloid leukemia, and may function as a leukemia disease gene, in part, through its interaction with BCL6. During hematopoietic cell differentiation, this gene is down-regulated. It is possibly involved in lymphoma pathogenesis since translocations associated with B-cell maligcies also deregulates its expression. Multiple transcript variants encoding several different isoforms have been found for this gene.Specifications
BCL11A | |
Polyclonal | |
Unconjugated | |
BCL11A | |
B cell CLL/lymphoma 11A, B cell lymphoma/leukemia 11A, BCL11A, COUP TF interacting protein 1, CTIP1, EVI 9, EVI9, HBFQTL5, KIAA1809, ZNF856 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
14025, 305589, 53335 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H165, Q9QYE3 | |
BCL11A | |
BCL11A Fusion Protein Ag2187 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.