Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL2L12 Antibody (SR1529), Novus Biologicals™
SDP

Catalog No. NB387900 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387900 100 μL
NB260367 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB387900 Supplier Novus Biologicals Supplier No. NBP322097100UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BCL2L12 Monoclonal antibody specifically detects BCL2L12 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation

Specifications

Antigen BCL2L12
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SR1529
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias Bcl-2 related proline-rich protein, bcl2-L-12, BCL2-like 12 (proline rich), bcl-2-like protein 12, Bcl-2-related proline-rich protein, BPR, MGC120313, MGC120314, MGC120315
Host Species Rabbit
Immunogen A synthesized peptide derived from human BCL2L12 (Uniprot number: Q9HB09)
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 83596
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.