Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL6B Antibody, Novus Biologicals™
SDP

Catalog No. NBP180434 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP180434 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP180434 Supplier Novus Biologicals Supplier No. NBP180434
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BCL6B Polyclonal specifically detects BCL6B in Human samples. It is validated for Western Blot.

Specifications

Antigen BCL6B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. NP_862827
Gene Alias BAZFBcl6-associated zinc finger protein, B-cell CLL/lymphoma 6 member B protein, B-cell CLL/lymphoma 6, member B, B-cell CLL/lymphoma 6, member B (zinc finger protein), ZBTB28, Zinc finger protein 62ZNF62
Gene Symbols BCL6B
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of human BCL6B. Peptide sequence SHVRIHTGEKPYHCDPCGLHFRHKSQLRLHLRQKHGAATNTKVHYHILGGP The peptide sequence for this immunogen was taken from within the described region.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 255877
Test Specificity Expected identity based on immunogen sequence: Bovine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Pig: 100%; Rabbit: 100%; Canine: 92%; Mouse: 92%; Rat: 92%.
Reconstitution Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Target Species Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.