Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL9-2 Antibody (1C9G2) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261705 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61705 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61705 Supplier Novus Biologicals Supplier No. NBP261705
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BCL9-2 Monoclonal specifically detects BCL9-2 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen BCL9-2
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 1C9G2
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Formulation PBS with 0.05% Sodium Azide
Gene Alias B-cell CLL/lymphoma 9-like, B-cell CLL/lymphoma 9-like protein, B-cell lymphoma 9-like protein, BCL9-like protein, DLNB11BCL9-2, nuclear co-factor of beta-catenin signalling, Protein BCL9-2
Gene Symbols BCL9L
Host Species Mouse
Immunogen Purified recombinant fragment of human BCL9L (AA: 606-751) expressed in E. Coli.
Molecular Weight of Antigen 157 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 283149
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.