Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCMA/TNFRSF17 Antibody (049), PerCP, Novus Biologicals™
SDP

Catalog No. NB350777 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB350777 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB350777 Supplier Novus Biologicals Supplier No. NBP312791PCP
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

BCMA/TNFRSF17 Monoclonal antibody specifically detects BCMA/TNFRSF17 in Human samples. It is validated for ELISA

Specifications

Antigen BCMA/TNFRSF17
Applications ELISA
Classification Monoclonal
Clone 049
Conjugate PerCP
Dilution ELISA
Formulation PBS
Gene Alias B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human BCMA/TNFRSF17 (Accession#: NP_001183.2; Met1-Ala54).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, Cytokine Research, Immunology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 608
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.