Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCMA/TNFRSF17 Antibody (Vicky-1), PerCP, Novus Biologicals™
SDP

Catalog No. NB002007 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002007 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002007 Supplier Novus Biologicals Supplier No. NBP197637PCP0.1ML
Add to Cart
Add to Cart

Rat Monoclonal Antibody

BCMA/TNFRSF17 Monoclonal specifically detects BCMA/TNFRSF17 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen BCMA/TNFRSF17
Applications Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Blocking Assay, Neutralization
Classification Monoclonal
Clone Vicky-1
Conjugate PerCP
Dilution Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Block/Neutralize
Formulation PBS
Gene Alias B cell maturation antigen, B-cell maturation protein, BCMAtumor necrosis factor receptor superfamily member 17, BCMB-cell maturation factor, CD269, CD269 antigen, tumor necrosis factor receptor superfamily, member 17
Gene Symbols TNFRSF17
Host Species Rat
Immunogen Recombinant human BCMA extracellular domain (aa 2-54).
Quantity 0.1 mL
Research Discipline Cytokine Research, Immunology
Gene ID (Entrez) 608
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.