Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCR/ABL Antibody (4C12), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB002665646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002665646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002665646 Supplier Novus Biologicals Supplier No. NB1002665JF646

Mouse Monoclonal Antibody

BCR/ABL Monoclonal specifically detects BCR/ABL in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen BCR/ABL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 4C12
Conjugate Janelia Fluor 646
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ABL, ALL, BCR ABL fusion protein, bcr/abl, BCR/ABL fusion protein, Bcr/c abl oncogene protein, BCR-ABL fusion protein, BCRe18/ABL1e3 fusion protein, breakpoint cluster region protein, c-ABL, CML, D22S662, JTK7, p150, PHL
Gene Symbols BCR
Host Species Mouse
Immunogen BCR/ABL peptide
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 613
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.