Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCR Antibody (1E11G12) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP261707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP2-61707 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP2-61707 Supplier Novus Biologicals Supplier No. NBP261707
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BCR Monoclonal specifically detects BCR in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen BCR
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 1E11G12
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000
Formulation PBS with 0.05% Sodium Azide
Gene Alias ALLD22S11, BCR1, breakpoint cluster region, breakpoint cluster region protein, CMLEC 2.7.11.1, D22S662, PHLFLJ16453, Renal carcinoma antigen NY-REN-26
Gene Symbols BCR
Host Species Mouse
Immunogen Purified recombinant fragment of human BCR (AA: 139-280) expressed in E. Coli.
Molecular Weight of Antigen 142.8 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Research Discipline Phospho Specific, Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 613
Target Species Human, Mouse
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.