Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCR, p Tyr276 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224460 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224460 0.02 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP224460 Supplier Novus Biologicals Supplier No. NBP224460
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

BCR Polyclonal specifically detects BCR in Human, Primate samples. It is validated for Western Blot.

Specifications

Antigen BCR (p Tyr276)
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 0.01-1.0 ug/ml
Formulation 0.1 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Alias ALLD22S11, BCR1, breakpoint cluster region, breakpoint cluster region protein, CMLEC 2.7.11.1, D22S662, PHLFLJ16453, Renal carcinoma antigen NY-REN-26
Gene Symbols BCR
Host Species Rabbit
Immunogen A portion of human phospho Bcr protein containing a phosphorylated tyrosine residue at position 276 was used as the immunogen for this phospho antibody.
Purification Method Affinity Purified
Quantity 0.02 mg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 613
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, monkey and mouse, 94% homologous in rat and cow, 89% homologous in chicken, 84% homologous in zebrafish and 81% homologous in xenopus.
Target Species Human, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.