Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BCS1L Mouse anti-Human, Mouse, Clone: 3G6H1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | BCS1L |
---|---|
Clone | 3G6H1 |
Concentration | 0.86 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6315
![]() ![]() |
Proteintech Group Inc
602121IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a homolog of the S. cerevisiae bcs1 protein which is involved in the assembly of complex III of the mitochondrial respiratory chain. The encoded protein does not contain a mitochondrial targeting sequence but experimental studies confirm that it is imported into mitochondria. Mutations in this gene are associated with mitochondrial complex III deficiency and the GRACILE syndrome. Five alternatively spliced transcripts encoding the same protein have been described.Specifications
BCS1L | |
0.86 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
BCS1L, BJS, FLNMS, GRACILE, h BCS, h BCS1, Hs.6719, Mitochondrial chaperone BCS1, PTD | |
BCS1L | |
IgG1 | |
Protein G | |
Antibody |
3G6H1 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
Q9CZP5, Q9Y276 | |
617, 66821 | |
BCS1L Fusion Protein Ag18280 | |
Primary | |
-20°C | |
BCS1L |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title