Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Annexin V-V450

Catalog No. bdb560506 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB560506 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB560506 Supplier BD Supplier No. 560506
Only null left
Add to Cart
Add to Cart

Serves as a sensitive probe for flow cytometric analysis of cells that are undergoing apoptosis

Identifies apoptosis at an earlier stage than assays based on nuclear changes such as DNA fragmentation.

  • Format: V450
  • Volume per Test: 5μL

Flow Cytometry

TRUSTED_SUSTAINABILITY

Specifications

Quantity 50 Tests
Sample Volume 5 μL
Storage Buffer Aqueous Buffered solution containing BSA and ≤0.09% Sodium Azide
Content And Storage 4°C
For Use With (Application) Flow Cytometry
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.