Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Bacto™ Dehydrated Culture Media: Tryptic Soy Broth (Soybean-Casein Digest Medium)
Shop All BD Diagnostic Systems Products
Click to view available options
:
Soy Broth (Soybean-Casein Digest Medium); 100g
Description
- General purpose medium used in qualitative procedures
- Conforms with USP and EP Specifications

Specifications
Specifications
Certifications/Compliance | AOAC, BAM, BS12, CCAM, CLSI, CMPH2, COMPF, EP, EPA, ISO, JP, MCM9, SMD, USDA, USP |
Content And Storage | 2°C to 25°C |
Form | Dehydrated |
Media Type | Tryptic Soy Broth (Soybean-Casein Digest Medium) |
Product Type | Microbiology Media |
Formulation | Pancreatic digest of Casein 17g/L, Papaic digest of Soybean 3g/L, Dextrose 2.5g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L |
For Use With (Application) | Cultivation of dermatophytes and other pathogenic and nonpathogenic fungi from clinical and nonclinical specimens |
Packaging Type | Bottle |
Quantity | 100 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction