Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: BiGGY Agar

Catalog No. b11027
Click to view available options
:
BiGGY Agar; 500g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used in detection, isolation and presumptive identification of Candida species.

  • Formulation: Bismuth Ammonium Citrate 5g/L, Sodium Sulfite 3g/L, Dextrose 10g/L, Glycine 10g/L, Yeast extract 1g/L, Agar 16g/L

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Form Dehydrated
Media Type BIGGY Agar
Product Type Agar
Formulation Bismuth Ammonium Citrate 5g/L, Sodium Sulfite 3g/L, Dextrose 10g/L, Glycine 10g/L, Yeast extract 1g/L, Agar 16g/L
For Use With (Application) For Candida
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.