Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Cary-Blair Transport Medium

Catalog No. b11102
Click to view available options
:
Cary-Blair Transport Medium; 500g
This item is not returnable. View return policy
This item is not returnable. View return policy

To collect, transport and preserve microbiological specimens, particularly Vibrio cholerae

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CCAM
Content And Storage 2°C to 8°C
Form Dehydrated
Media Type Cary-Blair Transport Medium
Product Type Cary-Blair Transport medium
Formulation Sodium Thioglycollate 1.5g/L, Disodium Phosphate 1.1g/L, Sodium Chloride 5g/L, Agar 5g/L
For Use With (Application) For collecting, transporting and preserving microbiological specimens, particularly Vibrio cholerae
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.