Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Columbia Agar Base

Catalog No. p-4901323 Shop All BD Diagnostic Systems Products
Change view
Click to view available options
Quantity:
500 g
2.26 kg
Packaging Type:
Bottle
Pail
For Use With (Application):
Growth of fastidious microbes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type For Use With (Application)
B11124 500 g Bottle Growth of fastidious microbes
B11125 2.26 kg Pail Growth of fastidious microbes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. B11124 Supplier BD Supplier No. 211124
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Used for cultivation of fastidious microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, COMPF, EP, ISO, JP, USP
Content And Storage 2 to 25°C
Form Dehydrated
Media Type Columbia Agar Base
Product Type Agar
Formulation Pancreatic digest of Casein 10g/L, meat Peptic digest 5g/L, Yeast extract 5g/L, heart Pancreatic digest 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 13.5g/L
For Use With (Application) Growth of fastidious microbes
Packaging Type Bottle
Quantity 500 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.