Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: DNase Test Agar
Click to view available options
:
DNase Test Agar; 500g
Specifications
Specifications
Certifications/Compliance | COMPF |
Content And Storage | Refrigerate |
Form | Dehydrated |
Media Type | DNase Test Agar |
Product Type | Agar |
Formulation | Pancreatic digest of Casein 15g/L, Papaic digest of Soybean Meal 5g/L, Deoxyribonucleic Acid 2g/L, Sodium Chloride 5g/L, Agar 15g/L |
For Use With (Application) | Detection of DNase activity to aid in the identification of bacteria isolated from clinical specimen |
Packaging Type | Bottle |
Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction