Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: EMB Agar Levine, without Lactose
Click to view available options
:
EMB Agar Levine without Lactose; 500g,
Description
- For genetic studies of enteric bacilli
- Formulation: Pancreatic digest of Gelatin 10g/L, Dipotassium Phosphate 2g/L, Eosin Y 0.4g/L, Methylene blue 65mg/L, Agar 15g/L
Certifications
AOAC, BAM, BS12, CCAM, CMPH2, COMPF, MCM9, SMD

Specifications
Specifications
Certifications/Compliance | AOAC, BAM, BS12, CCAM, CMPH2, COMPF, MCM9, SMD |
Form | Dehydrated |
Media Type | EMB Agar |
Product Type | Agar |
Formulation | Pancreatic digest of Gelatin 10g/L, Dipotassium Phosphate 2g/L, Eosin Y 0.4g/L, Methylene blue 65mg/L, Agar 15g/L |
For Use With (Application) | Isolation of gram negative enteric bacteria |
Packaging Type | Bottle |
Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction