Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Dehydrated Culture Media: Lysine Iron Agar

Catalog No. b11363
Click to view available options
:
Lysine Iron Agar; 500g
This item is not returnable. View return policy
This item is not returnable. View return policy

Differential medium to detect organisms that decarboxylate or deaminate lysine and produce hydrogen sulfide

Applications:

For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO, 9000, AOAC, BAM, CCAM, CMPH2, COMPF, MCM9, SMD, SMWW, USDA
Form Dehydrated
Media Type Lysine Iron Agar
Product Type Agar
Formulation Pancreatic digest of Gelatin 5g/L, Yeast extract 3g/L, Dextrose 1g/L, L-Lysine 10g/L, Ferric Ammonium Citrate 0.5g/L, Sodium Thiosulfate 0.04g/L, Bromcresol Purple 0.02g/L, Agar 13.5g/L
For Use With (Application) Isolation of Citrobacter freundii 8454, Escherichia coli 25922, Proteus vulgaris 9484, Providencia rustigianii 13159, Salmonella enterica subspecies Arizonae 13314, Salmonella enterica subspecies enterica serotype Paratyphi A 9150, Salmonella enterica subspecies enterica serotype Typhi 19430
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.