Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Mueller Hinton Broth
Click to view available options
:
Mueller Hinton Broth; 500g
Specifications
Specifications
Certifications/Compliance | CLSI |
Content And Storage | 2°C to 30°C |
Form | Dehydrated |
Media Type | Mueller Hinton Broth |
Product Type | Microbiology Media |
Formulation | Beef extract 3g/L, Acid Hydrolysate of Casein 17.5g/L, Starch 1.5g/L |
For Use With (Application) | Antimicrobial susceptibility testing of microorganisms by Broth dilution method |
Packaging Type | Bottle |
Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction