Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Phenylalanine Agar
Shop All BD Diagnostic Systems Products
Click to view available options
:
Phenylalanine Agar; 500g
Description
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.

Specifications
Specifications
Certifications/Compliance | BAM |
Content And Storage | 2°C to 8°C |
Form | Dehydrated |
Media Type | Phenylalanine Agar |
Product Type | Agar |
Formulation | DL-Phenylalanine 2g/L, Yeast extract 3g/L, Sodium Chloride 5g/L, Sodium Phosphate 1g/L, Agar 12g/L |
For Use With (Application) | Differentiation of Proteus and Providencia from other Enterobacteriaceae based on deamination of Phenylalanine |
Packaging Type | Bottle |
Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction