Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Purple broth base
Click to view available options
:
Purple broth base; 500g
Specifications
Specifications
Certifications/Compliance | ISO/DIS, 11014, AOAC, BAM, CCAM, COMPF, ISO, SMD, USDA |
Form | Dehydrated |
Media Type | Purple Broth Base |
Product Type | Microbiology Media |
Formulation | Pancreatic digest of Gelatin 10g/L, Sodium Chloride 5g/L, Bromcresol Purple 0.02g/L |
For Use With (Application) | Isolation of gram negative enteric bacilli |
Packaging Type | Bottle |
Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction