Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Simmons Citrate Agar
Shop All BD Diagnostic Systems Products
Click to view available options
:
Simmons Citrate Agar; 500g
Specifications
Specifications
| Certifications/Compliance | CE, AOAC, BAM, CCAM, COMPF, ISO |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Simmons Citrate Agar |
| Product Type | Agar |
| Formulation | Ammonium Dihydrogen Phosphate 1g/L, Dipotassium Phosphate 1g/L, Sodium Chloride 5g/L, Sodium Citrate 2g/L, Magnesium Sulfate 0.2g/L, Agar 15g/L, Bromthymol blue 0.08g/L |
| For Use With (Application) | For differentiation of gram negative bacteria on the basis of citrate utilization |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction