Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD BBL™ Trypticase™ Soy Broth (Soybean-Casein Digest Medium)

Catalog No. p-4869070
Click to view available options
Quantity:
500 g
5 lb.
25 lb.
Packaging Type:
Bottle
Pail
This item is not returnable. View return policy
This item is not returnable. View return policy

Conforms with USP and EP specifications. BD™ BBL™ Trypticase™ Soy Broth (Soybean-Casein Digest Medium) is a dehydrated, general purpose media for isolation, detection and cultivation of wide variety of microorganisms.

Trypticase Soy Broth (Soybean-Casein Digest Medium) is a general purpose medium used in qualitative procedures for the cultivation of fastidious and non-fastidious microorganisms from a variety of clinical and nonclinical specimens. TSB is a nutritious medium that will support the growth of a wide variety of microorganisms, including common aerobic, facultative and anaerobic bacteria and fungi.

  • This formulation is included in the USP as a medium for use in performing microbial enumeration tests and tests for specified microorganisms when testing nonsterile pharmaceutical products.
  • TSB was chosen by the USDA Animal and Plant Health Inspection Service for detecting viable bacteria in live vaccines.
  • TSB is recommended for testing bacterial contaminants in cosmetics and complies with established standards in the food industry.
  • Because of its capacity for growth promotion, TSB is also recommended for use as the inoculum broth for disc diffusion and agar dilution antimicrobial susceptibility testing as standardized by the Clinical and Laboratory Standards Institute (CLSI).
  • Used in the isolation of: Aspergillus brasiliensis (niger) 16404, Bacillus subtilis 6633, Candida albicans 10231, Escherichia coli 8739, Pseudomonas aeruginosa 9027, Salmonella enterica subspecies enterica serotype Typhimurium 14028, Staphylococcus aureus 6538

Certifications

AOAC, BAM, BS12, CCAM, CLSI, CMPH2, COMPF, EP, EPA, ISO, JP, MCM9, SMD, USDA, USP

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, BS12, CCAM, CLSI, CMPH2, COMPF, EP, EPA, ISO, JP, MCM9, SMD, USDA, USP
Color Light tan to yellow (prepared)
Content And Storage 2 to 25°C
Form Dehydrated (free-flowing, homogenous)
Media Type Trypticase Soy Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 17g/L, Papaic digest of Soybean 3g/L, Sodium Chloride 5g/L, Dipotassium Phosphate 2.5g/L, Dextrose 2.5g/L
For Use With (Application) Cultivation of fastidious and non-fastidious microorganisms from a variety of clinical and nonclinical specimens
Packaging Type Pail
Quantity 25 lb.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.