Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD BBL™ Dehydrated Culture Media: Tryptone Glucose Yeast Agar (Standard Methods Agar)
Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
500 g
2268 g
Packaging Type:
Bottle
Pail
Specifications
Specifications
| Certifications/Compliance | CE, AOAC, BAM, CCAM, COMPF, EPA, ISO, SMD, SMWW, USDA |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Standard Methods Agar (Tryptone Glucose Yeast Agar) |
| Product Type | Agar |
| Formulation | Pancreatic digest of Casein 5g/L, Yeast extract 2.5g/L, Dextrose 1g/L, Agar 15g/L |
| For Use With (Application) | For obtaining microbial plate counts from Milk and dairy product, food Water and other materials of sanitary importance |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction