Learn More
Description
BrdU in culture is incorporated into the DNA during DNA synthesis. Cellular incorporation of BrdU can be detected by anti-BrdU specific antibodies following membrane permeabilization through flow cytometry or immunohistochemistry.
- Free of preservatives; handle under aseptic conditions
- Provided as five vials, each containing 0.5mL of a 10mg/mL solution diluted in 1X DPBS
Specifications
Specifications
| Format | Liquid |
| pH | 7.2 |
| Product Type | Bromodeoxyuridine (BrdU) |
| Content And Storage | -80°C |
| Concentration | 10 mg/mL |
| Quantity | 25 mg |
| For Use With (Application) | Cellular incorporation of BrdU can be detected by anti-BrdU specific Antibodies following membrane permeabilization through flow cytometry or immunohistochemistry |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.