Learn More
Description
The bead mixture contains bead positions A4, A5, A6, A7, A8, A9, B4, B5, B6, B7, B8, B9, C4, C5, C6, C7, C8, C9, D4, D5, D6, D7, D8, D9, E4, E5, E6, E7, E8 and E9. It is only a guarantee that the instrument and FCAP Array Software (Cat. No. 641488) can analyze the 30 plex correctly
- 30 Plex Bead Mixture is a cocktail
- Contains 30 bead populations from BD CBA Flex Set
- Has distinct fluorescence intensities resolvable in NIR and Red channels of BD FACSArray bioanalyzer
- Storage Buffer: Aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide
Array
Specifications
Specifications
| Certifications/Compliance | RUO |
| Content And Storage | 4°C |
| For Use With (Equipment) | Flow cytometer |
| No. of Tests | 10 |
| Includes | Aqueous buffered solution containing fetal bovine serum and ≤0.09% sodium azide |
| Quantity | 30 bead |
| Volume (Metric) | 25 μL |
| For Use With (Application) | Flow Cytometry Calibration and Alignment |
Research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.