Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
BD Dehydrated Media M17 Agar
Shop All BD Diagnostic Systems Products
                    Click to view available options
                
                
                    
                            
                                :
                            
                        
                        
                            
                                500g
                            
                        
                    Specifications
Specifications
| Content And Storage | <30°C | 
| Form | Dehydrated | 
| Product Type | Agar | 
| Formulation | Pancreatic digest of Casein 5g/L, Soy Peptone 5g/L, Beef extract 5g/L, Yeast extract 2.5g/L, Ascorbic Acid 0.5g/L, Magnesium Sulfate 0.25g/L, Disodium-beta-glycerophosphate 19g/L, Agar 11g/L | 
| For Use With (Application) | strains used in molecular microbiology procedures | 
| Packaging Type | Bottle | 
| Quantity | 500 g | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
             
                                