Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Antisera for Serological Confirmation of E. coli O157:H7

E. ColiO Antiserum O157 and E. Coli H Antiserum H7 are used for identifying Escherichia coli O157:H7

Supplier:  BD 221591

 View more versions of this product

Catalog No. D215947

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Description

Description

E. coli O157:H7 is an enteric pathogen that requires only a low inoculum to cause disease. Transmission is usually via food items. Serotyping of the entero-hemorrhagic E. coli is useful in the epidemiological documentation of the spread of a particular strain in a foodborne outbreak. At the time of use, test both positive and negative control cultures to check performance of the antiserum.

  • Product type: antigens and antisera
  • Groupings: D through K Products
  • Type: antisera
  • Sub type: E.coli - Lyophilized, re-hydrate to 3mL
  • Shelf Life: 1095 days
  • Label has Natural Rubber Latex statement

Certifications

CE marked

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CE Marked
Refrigerate, store at 2°C to 8°C
Antiserum
For tube test for serological confirmation of E. coli O157:H7
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 1095 Days

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.