Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Antisera for Serological Confirmation of E. coli O157:H7

E. ColiO Antiserum O157 and E. Coli H Antiserum H7 are used for identifying Escherichia coli O157:H7

$306.00

Specifications

Certifications/Compliance CE Marked
Content And Storage Refrigerate, store at 2°C to 8°C
Product Type Antiserum
For Use With (Application) For tube test for serological confirmation of E. coli O157:H7
View More Specs

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
DF2970-47-7
View Documents
BD
229701
Antiserum
Each for $306.00
Only null left
Add to Cart
 
D215947
View Documents
BD
221591
Antiserum N/A N/A N/A
Description

Description

E. coli O157:H7 is an enteric pathogen that requires only a low inoculum to cause disease. Transmission is usually via food items. Serotyping of the entero-hemorrhagic E. coli is useful in the epidemiological documentation of the spread of a particular strain in a foodborne outbreak. At the time of use, test both positive and negative control cultures to check performance of the antiserum.

  • Product type: antigens and antisera
  • Groupings: D through K Products
  • Type: antisera
  • Sub type: E.coli - Lyophilized, re-hydrate to 3mL
  • Shelf Life: 1095 days
  • Label has Natural Rubber Latex statement

Certifications

CE marked

Specifications

Specifications

CE Marked
Refrigerate, store at 2°C to 8°C
Antiserum
For tube test for serological confirmation of E. coli O157:H7
Videos
Safety and Handling

Safety and Handling

ShelfLife : 1095 Days

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.