Learn More
Description
- Penassay Seed Agar Microbiological assay of antibiotics
Specifications
Specifications
| Certifications/Compliance | AOAC, EP, USP |
| Content And Storage | 2°C to 8°C |
| Form | Dehydrated |
| Media Type | Antibiotic Medium 1 |
| Product Type | Antibiotic Medium |
| Formulation | Beef extract 1.5g/L, Yeast extract 3g/L, Pancreatic digest of Casein 4g/L, Peptone 6g/L, Dextrose 1g/L, Agar 15g/L |
| For Use With (Application) | Penassay Seed Agar Microbiological assay of Antibiotics |
| Packaging Type | Bottle |
| Quantity | 500 g |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.