Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Baird-Parker Agar Base

Catalog No. p-4902038 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
This item is not returnable. View return policy
This item is not returnable. View return policy

Use with EY Tellurite Enrichment for the detection and enumeration of coagulase positive staphylococci in foods and other materials

  • Formulation: Pancreatic digest of Casein 10g/L, Beef extract 5g/L, Yeast extract 1g/L, Glycine 12g/L, Sodium Pyruvate 10g/L, Lithium Chloride 5g/L, Agar 20g/L

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage <30°C
Form Dehydrated
Media Type Baird-Parker Agar Base
Product Type Agar
Formulation Pancreatic digest of Casein 10g/L, Beef extract 5g/L, Yeast extract 1g/L, Glycine 12g/L, Sodium Pyruvate 10g/L, Lithium Chloride 5g/L, Agar 20g/L
For Use With (Application) For coagulase positive Staphylococcus
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.