Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: BCG Dextrose Agar
Shop All BD Diagnostic Systems Products
Click to view available options
:
Snyder Test Agar (BCG Dextrose Agar); 500g
Description
- Also known as Snyder Test Agar
Specifications
Specifications
| Certifications/Compliance | CE |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Snyder Test Agar |
| Product Type | Agar |
| Formulation | Proteose Peptone 10g/L, Pancreatic digest of Casein 10g/L, Dextrose 20g/L, Sodium Chloride 5g/L, Agar 20g/L, Bromcresol green 0.02g/L |
| For Use With (Application) | For estimating the relative number of lactoBacilli in saliva based on Acid production |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction