Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Brilliant Green Bile Agar

Catalog No. DF0014175
Click to view available options
:
Brilliant Green Bile Agar; 500g
This item is not returnable. View return policy
This item is not returnable. View return policy

For selective differentiation of coliform bacteria

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance COMPF
Content And Storage <30°C
Form Dehydrated
Media Type Brilliant Green Bile Agar
Product Type Agar
Formulation Peptone 8.25g/L, Lactose 1.9g/L, Oxgall 2.95mg/L, Sodium Sulfite 205mg/L, Ferric Chloride 29.5mg/L, Monopotassium Phosphate 15.3mg/L, Agar 10.15g/L, Erioglaucine 64.9mg/L, Basic Fuchsin 77.6mg/L, Brilliant green 29.5μg/L
For Use With (Application) Selective differentiation of Coliform bacteria
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.