Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Buffered Glucose Broth
Shop All BD Diagnostic Systems Products
Click to view available options
:
MR-VP Medium (Buffered Glucose Broth); 500g
Specifications
Specifications
Certifications/Compliance | ISO, 9000, AOAC, BAM, CCAM, COMPF, ISO, SMD, SMWW, USDA |
Content And Storage | <30°C |
Form | Dehydrated |
Media Type | MR-VP Medium |
Product Type | Microbiology Media |
Formulation | Buffered Peptone 7g/L, Dipotassium Phosphate 5g/L, Dextrose 5g/L |
For Use With (Application) | Isolation of Enterbacter aerogenes 13048, Escherichia coli 25922, Klebsiella pneumoniae subspecies pneumoniae 23357 |
Packaging Type | Bottle |
Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction