Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: CIN Agar Base, Modified
Shop All BD Diagnostic Systems Products
Click to view available options
:
Yersinia Selective Agar Base (CIN Agar Base, Modified); 500g
Description
Order Info
Prices are for units of one each unless otherwise specified.
Specifications
Specifications
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Yersinia selective Agar base (CIN Agar base, modified) |
| Product Type | Agar |
| Formulation | Peptone 17g/L, Proteose Peptone 3g/L, Yeast extract 2g/L, Mannitol 20g/L, Sodium Pyruvate 2g/L, Sodium Chloride 1g/L, Magnesium Sulfate Heptahydrate 10mg/L, Sodium Desoxycholate 0.5g/L, Sodium Cholate 0.5g/L, Irgasan™ 4mg/L, Agar 13.5g/L |
| For Use With (Application) | Isolation of Yersinia entero-colitica from a variety of clinical and nonclinical specimens |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction