Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Columbia Blood Agar Base
Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Specifications
Specifications
Certifications/Compliance | AOAC, BAM, EP, ISO |
Content And Storage | <30°C |
Form | Dehydrated |
Media Type | Columbia Blood Agar Base |
Product Type | Agar |
Formulation | Pancreatic digest of Casein 10g/L, Proteose Peptone No. 3 5g/L, Yeast extract 5g/L, Beef heart infusion from 500 g 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 15g/L |
For Use With (Application) | For enhanced beta-hemolytic reactions after overnight incubation and for cultivation of fastidious microorganisms |
Packaging Type | Pail |
Quantity | 2 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction