Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Columbia Blood Agar Base

Catalog No. p-4901432 Shop All BD Diagnostic Systems Products
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
This item is not returnable. View return policy

Catalog No. DF0792-17-3

Supplier: BD 279240

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

For enhanced beta-hemolytic reactions after overnight incubation and for cultivation of fastidious microorganisms

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance AOAC, BAM, EP, ISO
Content And Storage <30°C
Form Dehydrated
Media Type Columbia Blood Agar Base
Product Type Agar
Formulation Pancreatic digest of Casein 10g/L, Proteose Peptone No. 3 5g/L, Yeast extract 5g/L, Beef heart infusion from 500 g 3g/L, Corn Starch 1g/L, Sodium Chloride 5g/L, Agar 15g/L
For Use With (Application) For enhanced beta-hemolytic reactions after overnight incubation and for cultivation of fastidious microorganisms
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.