Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Dextrose Tryptone Agar

Catalog No. df0080174 Shop All BD Diagnostic Systems Products
Click to view available options
:
Dextrose Tryptone Agar; 500g
This item is not returnable. View return policy
This item is not returnable. View return policy

For cultivation of thermophilic flat-sour microorganisms associated with food spoilage

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance COMPF
Content And Storage Refrigerate
Form Dehydrated
Media Type Dextrose Tryptone Agar
Product Type Agar
Formulation Tryptone 8g/L, Peptone 2g/L, Dextrose 5g/L, Agar 15g/L, Bromcresol Purple 0.04g/L
For Use With (Application) Cultivation of thermophilic “flat-sour” microorganisms associated with food spoilage
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.