Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: MacConkey Sorbitol Agar

Catalog No. df0079177
Click to view available options
:
MacConkey Sorbitol Agar; 500g
This item is not returnable. View return policy
This item is not returnable. View return policy

For isolation and differentiation of enteropathogenic E. coli serotypes

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

Certifications

CE, BAM, BS12, CCAM, CMPH2, COMPF, ISO, MCM9, SMWW

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE, BAM, BS12, CCAM, CMPH2, COMPF, ISO, MCM9, SMWW
Content And Storage Refrigerate
Form Dehydrated
Media Type MacConkey Sorbitol Agar
Product Type Agar
Formulation Peptone 15.5g/L, Proteose Peptone 3g/L, D-Sorbitol 10g/L, Bile Salts 1.5g/L, Sodium Chloride 5g/L, Agar 15g/L, Neutral red 0.03g/L, Crystal Violet 1mg/L
For Use With (Application) Isolation and differentiation of enteric pathogenic escherichia coli Serotype
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.