Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
BD Difco™ Malt Extract Broth
Shop All BD Diagnostic Systems Products
                    Click to view available options
                
                
                    
                            
                                :
                            
                        
                        
                            
                                Malt Extract Broth; 500g
                            
                        
                    Description
The use of malt and malt extracts for the propagation of yeasts and molds is quite common. Malt Extract Broth is recommended for the examination of yeasts and molds in the U.S. Food and Drug Administration’s Bacteriological Analytical Manual.
- Contains malt extract which provides the carbon, protein, and nutrient sources required for growth of microorganisms.
- Maltose is added as an energy source, and Dextrose is included as a source of fermentable carbohydrate.
- Yeast extract provides the vitamins and cofactors required for growth and additional sources of nitrogen and carbon.
- Provides good recovery of Aspergillus brasiliensis (niger), Candida albicans and Saccharomyces cerevisiae.
Order Info
See our web site or the Fisher Scientific™ HealthCare catalog for product information.
Certifications
CE, BAM
 
            Specifications
Specifications
| Certifications/Compliance | CE, BAM | 
| Color | Light beige | 
| Content And Storage | Refrigerate | 
| Form | Dehydrated | 
| Media Type | Malt Extract Broth | 
| Product Type | Microbiology Media | 
| Formulation | Malt extract 6g/L, Maltose, Technical 1.8g/L, Dextrose 6g/L, Yeast extract 1.2g/L | 
| For Use With (Application) | For Isolation, detection and enumeration of yeasts and molds | 
| Packaging Type | Bottle | 
| Quantity | 500 g | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            