Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Mycological Agar
Click to view available options
:
Mycological Agar; 500g
Specifications
Specifications
Certifications/Compliance | CE |
Content And Storage | Refrigerate |
Form | Dehydrated |
Media Type | Mycological Agar |
Product Type | Agar |
Formulation | Soy Peptone 10g/L, Dextrose 10g/L, Agar 15g/L |
For Use With (Application) | Cultivation and maintenance of fungi, for the demonstration of chromogenesis and for obtaining Yeast and mold count |
Packaging Type | Bottle |
Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction