Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Pseudomonas Agar F

Catalog No. df0448171 Shop All BD Diagnostic Systems Products
Click to view available options
:
Pseudomonas Agar F; 500g

Specifications

Certifications/Compliance BAM, CCAM
Content And Storage <30°C
Form Dehydrated
Media Type Pseudomonas Agar F
Product Type Agar
Formulation Pancreatic digest of Casein 10g/L, Proteose Peptone No. 3 10g/L, Dipotassium Phosphate 1.5g/L, Magnesium Sulfate 1.5g/L, Agar 15g/L
For Use With (Application) For enhancement of Pyocyanin production by Pseudomonas
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.