Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Raka-Ray No. 3 Medium
Shop All BD Diagnostic Systems Products
Click to view available options
:
Raka-Ray No. 3; 500g
Description
- Formulation: Yeast extract 5g/L, Tryptone 20g/L, liver Concentrate 1g/L, Maltose 10g/L, Fructose 5g/L, Glucose 5g/L, Betaine Hydrochloride 2g/L, Diammonium Citrate 2g/L, Potassium Aspartate 2.5g/L, Magnesium Sulfate 0.98g/L, Manganese Sulfate 0.42g/L, Dipotassium Phosphate 2g, N-Acetyl Glucosamine 0.5g, Potassium Glutamate 2.5g, Agar 16g
Order Info
Prices are for units of one each unless otherwise specified.
Specifications
Specifications
| Content And Storage | <30°C |
| Form | Dehydrated |
| Media Type | Raka-Ray No. 3 |
| Product Type | Dehydrated Culture Media |
| Formulation | Yeast extract 5g/L, Tryptone 20g/L, liver Concentrate 1g/L, Maltose 10g/L, Fructose 5g/L, Glucose 5g/L, Betaine Hydrochloride 2g/L, Diammonium Citrate 2g/L, Potassium Aspartate 2.5g/L, Magnesium Sulfate 0.98g/L, Manganese Sulfate 0.42g/L, Dipotassium Phosphate 2g/L, N-Acetyl Glucosamine 0.5g/L, Potassium Glutamate 2.5g/L, Agar 16g/L |
| For Use With (Application) | For enumerating heterotrophic organisms in treated potable Water |
| Packaging Type | Bottle |
| Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction