Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BD Difco™ Dehydrated Culture Media: Rappaport Vassiliadis R10 Broth

Catalog No. d185817 Shop All BD Diagnostic Systems Products
Click to view available options
:
Rappaport-Vassiliadis; 500g
This item is not returnable. View return policy
This item is not returnable. View return policy

For selectively enriching Salmonella from meat and dairy products, feces and sewage polluted water

  • Formulation: Pancreatic digest of Casein 4.54g/L, Sodium Chloride 7.2g/L, Monopotassium Phosphate 1.45g/L, Magnesium Chloride (anhydrous) 13.4g/L, Malachite green Oxalate 36mg/L

Order Info

Prices are for units of one each unless otherwise specified.

Certifications

IDF, ISO, USDA

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance IDF, ISO, USDA
Content And Storage <30°C
Form Dehydrated
Media Type Rappaport-Vassiliadis R10 Broth
Product Type Microbiology Media
Formulation Pancreatic digest of Casein 4.54g/L, Sodium Chloride 7.2g/L, Monopotassium Phosphate 1.45g/L, Magnesium Chloride (anhydrous) 13.4g/L, Malachite green Oxalate 36mg/L
For Use With (Application) Selective enrichment of Salmonella from food, polluted Water and other materials, selective media recommended by the European Chocolate Manufacturer's Association and the AOAC for rapid detection of motile Salmonella
Packaging Type Bottle
Quantity 500 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.