Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Staphylococcus Medium 110
Click to view available options
:
Staphylococcus; Medium 110; 500g
Description
- Also known as Stone Gelatin Agar

Specifications
Specifications
Certifications/Compliance | ISO, 9000 |
Content And Storage | <30°C |
Form | Dehydrated |
Media Type | Staphylococcus Medium 110 |
Product Type | Staphylococcus Medium |
Formulation | Pancreatic digest of Casein 10g/L, Yeast extract 2.5g/L, Gelatin 30g/L, Lactose 2g/L, D-Mannitol 10g/L, Sodium Chloride 75g/L, Dipotassium Phosphate 5g/L, Agar 15g/L |
For Use With (Application) | Isolation of Escherichia coli 25922, Staphylococcus aureus 25923, Staphylococcus epidermidis 12228 |
Packaging Type | Bottle |
Quantity | 500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction